Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromic-Sulfuric Acid Cleaning Solution for Cleaning Laboratory Glassware, Reagents

Catalog No. p-200021811
Change view
Click to view available options
Quantity:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-702 1 L
01-335-701 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-335-702 Supplier Reagents Holdings Llc Supplier No. CC0048001C1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chromic-Sulfuric Acid Cleaning Solution for Cleaning Laboratory Glassware, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Color-coded No
Form Liquid
pH 0
Sterility Non-sterile
Autoclavable Not Autoclavable
Boiling Point 260°C
Color Brown
Packaging Safety Coated Glass
Quantity 1 L
Solubility Completely miscible
Volume (Metric) 1 L
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.