Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cleaning Solution, Chromic-Sulfuric Acid Solution, Ricca Chemical
SDP

Catalog No. 21501 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Volume (Metric):
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
21501 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21501 Supplier Ricca Chemical Company Supplier No. 21501
Add to Cart
Add to Cart

Chromic-Sulfuric Acid Solution is a cleaning solution for cleaning laboratory glassware.

  • Clear, brown liquid
  • 7789-12-0 (Sodium Dichromate Dihydrate), 1333-82-0 (Chromium (VI) Trioxide), 7664-93-9 (Sulfuric Acid)

Specifications

Product Type Cleaning Solution
Color-coded No
Form Liquid
Scent Unscented
Sterility Non-sterile
Appearance Clear Brown
Color Brown
Content And Storage Corrosive
Format Solution
Volume (Metric) 4 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.