Learn More
Aqua Solutions Chromium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
SDP
Catalog No. NC1781912
Encompass
CAS 7789-09-5 NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet. ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material. Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation.
Catalog No. NC1781912 Supplier Aqua Solutions Supplier No. SXPLCR22X
Add to Cart
Add to Cart
CAS 7789-09-5 NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet. ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material. Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation.