Learn More
Revvity Health Sciences Inc Chromium-51 Radionuclide, 1mCi, EDTA Complex in 0.005M EDTA (pH 7.0)
Catalog No. 502812377
Encompass
Chromium-51 Radionuclide, 1mCi, EDTA Complex in 0.005M EDTA (pH 7.0)
Catalog No. 50-281-2377 Supplier Revvity Health Sciences Inc Supplier No. NEZ147001MC
Chromium-51 Radionuclide, 1mCi, EDTA Complex in 0.005M EDTA (pH 7.0)