Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium arsenide, 99% (metals basis), Thermo Scientific™

Catalog No. AA3920506
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3920506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3920506 Supplier Thermo Scientific Chemicals Supplier No. 3920506

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12254-85-2
Molecular Formula AsCr
Molecular Weight (g/mol) 126.92
MDL Number MFCD00198045
InChI Key OAMWSOUCVTVANH-UHFFFAOYSA-N
Synonym chromiumylidynearsane, chromium arsenide

Specifications

Physical Form Powder
Quantity 5 g
UN Number UN1557
Formula Weight 178.98
Percent Purity 99%
Assay (metals basis)
Chemical Name or Material Chromium arsenide
Hazard Category Danger
Hazard Statement GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
Precautionary Statement GHS P Statement
P261-P301+P310-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.