Learn More
Transene Inc Chromium Etchant 1020, Quart
SDP
Catalog No. NC0276919
Encompass
Transene Chromium Etchants 1020 and 1020AC are high purity ceric ammonium nitrate systems for precise, clean etching of chromium and chromium oxide films. Chromium Etchant 1020AC is nitric-acid free and exhibits a slower etch rate. Both etchants are compatible with positive and negative photoresists. Chromium Etchant 1020 is filtered to remove all particulates above 0.2 microns. Minimal undercutting is achieved with Chromium Etchant 1020AC.
This item is not returnable. View return policy
Catalog No. NC0276919 Supplier Transene Inc Supplier No. 0600021000
Transene Chromium Etchants 1020 and 1020AC are high purity ceric ammonium nitrate systems for precise, clean etching of chromium and chromium oxide films. Chromium Etchant 1020AC is nitric-acid free and exhibits a slower etch rate. Both etchants are compatible with positive and negative photoresists. Chromium Etchant 1020 is filtered to remove all particulates above 0.2 microns. Minimal undercutting is achieved with Chromium Etchant 1020AC.