Learn More
Strem, An Ascensus Company CAS# 7440-47-3. 100g. Chromium flakes (99.3%). MFCD00010944
SDP
Catalog No. 5090114572
Encompass
CAS# 7440-47-3. 100g. Chromium flakes (99.3%). MFCD00010944. Molecular Weight: 52.01. Molecular Formula: Cr. Color/form: 19 mm and down Strem# 93-2456. http://www.strem.com/catalog/v/93-2456/
Catalog No. 50-901-14572 Supplier Strem, An Ascensus Company Supplier No. 932456100G
CAS# 7440-47-3. 100g. Chromium flakes (99.3%). MFCD00010944. Molecular Weight: 52.01. Molecular Formula: Cr. Color/form: 19 mm and down Strem# 93-2456. http://www.strem.com/catalog/v/93-2456/