Learn More
Strem, An Ascensus Company CAS# 10049-05-5. 25g. Chromium(II) chloride, anhydrous (99.9%-Cr). MFCD00010947
SDP
Catalog No. 502016675
Encompass
CAS# 10049-05-5. 25g. Chromium(II) chloride, anhydrous (99.9%-Cr). MFCD00010947. Molecular Weight: 122.90. Molecular Formula: CrCl2. Color/form: off-white to pale green pwdr. Strem# 93-2405. http://www.strem.com/catalog/v/93-2405/
Catalog No. 50-201-6675 Supplier Strem, An Ascensus Company Supplier No. 93240525G
CAS# 10049-05-5. 25g. Chromium(II) chloride, anhydrous (99.9%-Cr). MFCD00010947. Molecular Weight: 122.90. Molecular Formula: CrCl2. Color/form: off-white to pale green pwdr. Strem# 93-2405. http://www.strem.com/catalog/v/93-2405/