Learn More
Strem, An Ascensus Company CAS# 39430-51-8. 250g. Chromium(III) acetate, basic. MFCD00058912
SDP
Catalog No. 5090114531
Encompass
CAS# 39430-51-8. 250g. Chromium(III) acetate, basic. MFCD00058912. Molecular Weight: 603.32. Molecular Formula: Cr3(OH)2(OOCCH3)7. Color/form: blue to green pwdr. Strem# 93-2401. http://www.strem.com/catalog/v/93-2401/
Catalog No. 50-901-14531 Supplier Strem, An Ascensus Company Supplier No. 932401250G
CAS# 39430-51-8. 250g. Chromium(III) acetate, basic. MFCD00058912. Molecular Weight: 603.32. Molecular Formula: Cr3(OH)2(OOCCH3)7. Color/form: blue to green pwdr. Strem# 93-2401. http://www.strem.com/catalog/v/93-2401/