Learn More
Strem, An Ascensus Company CAS# 21679-31-2. 250g. Chromium(III) acetylacetonate, 97.5%. MFCD00000015
SDP
Catalog No. 5090110857
Encompass
CAS# 21679-31-2. 250g. Chromium(III) acetylacetonate, 97.5%. MFCD00000015. Molecular Weight: 349.33. Molecular Formula: Cr(CH3COCHCOCH3)3. Color/form: maroon xtl. Strem# 24-0160. http://www.strem.com/catalog/v/24-0160/
Catalog No. 50-901-10857 Supplier Strem, An Ascensus Company Supplier No. 240160250G
CAS# 21679-31-2. 250g. Chromium(III) acetylacetonate, 97.5%. MFCD00000015. Molecular Weight: 349.33. Molecular Formula: Cr(CH3COCHCOCH3)3. Color/form: maroon xtl. Strem# 24-0160. http://www.strem.com/catalog/v/24-0160/