Learn More
Strem, An Ascensus Company CAS# 13478-06-3. 25g. Chromium(III) bromide hexahydrate. MFCD00150214
SDP
Catalog No. 5090114544
Encompass
CAS# 13478-06-3. 25g. Chromium(III) bromide hexahydrate. MFCD00150214. Molecular Weight: 291.69 (399.81). Molecular Formula: CrBr3∙6H2O. Color/form: green xtl. Strem# 93-2415. http://www.strem.com/catalog/v/93-2415/
Catalog No. 50-901-14544 Supplier Strem, An Ascensus Company Supplier No. 93241525G
CAS# 13478-06-3. 25g. Chromium(III) bromide hexahydrate. MFCD00150214. Molecular Weight: 291.69 (399.81). Molecular Formula: CrBr3∙6H2O. Color/form: green xtl. Strem# 93-2415. http://www.strem.com/catalog/v/93-2415/