Learn More
Strem, An Ascensus Company CAS# 10025-73-7. 10g. Chromium(III) chloride, anhydrous (99.9%-Cr). MFCD00010948
SDP
Catalog No. 5090114566
Encompass
CAS# 10025-73-7. 10g. Chromium(III) chloride, anhydrous (99.9%-Cr). MFCD00010948. Molecular Weight: 158.35. Molecular Formula: CrCl3. Color/form: purple flakes (< 1/8'') Strem# 93-2442. http://www.strem.com/catalog/v/93-2442/
Catalog No. 50-901-14566 Supplier Strem, An Ascensus Company Supplier No. 93244210G
CAS# 10025-73-7. 10g. Chromium(III) chloride, anhydrous (99.9%-Cr). MFCD00010948. Molecular Weight: 158.35. Molecular Formula: CrCl3. Color/form: purple flakes (< 1/8'') Strem# 93-2442. http://www.strem.com/catalog/v/93-2442/