Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium(III) chloride, anhydrous, 98%

Catalog No. p-7048978
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1233618 50 g
AA1233630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1233618 Supplier Thermo Scientific Chemicals Supplier No. 01233618
Only null left
Add to Cart

CAS: 10025-73-7

Chromium chloride and chromium sulfate have been used as dietary supplements. Chromium(III) chloride and chromium(III) sulfate can be used in the manufacture of food for particular nutritional uses and in food supplements. It is found in miscellaneous applications, such as catalysts, hardeners, mordents, crosslinking and binding agents. It is used to make very pure chromium and other chromium compounds, as a mordant in dyeing cotton, and in chromizing metals. It is too used for trivalent electroplating .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-73-7
MDL Number MFCD00010948
Synonym chromium iii chloride, chromium 3+ trichloride, anhydrous chromium iii chloride, basic chromic chloride, chromium chloride, basic, chromium trichl, chromium iii ; chloride, acmc-1ai6u, ksc492k0f

Specifications

Boiling Point 1300°C (sublimation)
Color Violet
Physical Form Crystalline
Quantity 50 g
UN Number UN3260
Merck Index 14,2222
Solubility Information Partially soluble in water.
Formula Weight 158.35
Percent Purity 98%
Sensitivity Hygroscopic
Density 2.87 g/mL
Chemical Name or Material Chromium(III) chloride, Anhydrous
Show More Show Less
Hazard Category H290-H302-H317
Hazard Statement GHS H Statement
H334-H314-H318-H302-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P234-P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P390-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 233-038-3
RTECSNumber GB5425000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.