Learn More
Chromium(III) chloride, anhydrous, 98%
Description
Chromium chloride and chromium sulfate have been used as dietary supplements. Chromium(III) chloride and chromium(III) sulfate can be used in the manufacture of food for particular nutritional uses and in food supplements. It is found in miscellaneous applications, such as catalysts, hardeners, mordents, crosslinking and binding agents. It is used to make very pure chromium and other chromium compounds, as a mordant in dyeing cotton, and in chromizing metals. It is too used for trivalent electroplating .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 1300°C (sublimation) |
Color | Violet |
Physical Form | Crystalline |
Quantity | 50 g |
UN Number | UN3260 |
Merck Index | 14,2222 |
Solubility Information | Partially soluble in water. |
Formula Weight | 158.35 |
Percent Purity | 98% |
Sensitivity | Hygroscopic |
Show More |
Safety and Handling
Hazard Category | H290-H302-H317 |
Hazard Statement | GHS H Statement H334-H314-H318-H302-H317 May cause allergy or asthma symptoms or breathing difficulties if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. May cause an allergic skin reaction. |
Precautionary Statement | P234-P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P390-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S. |
EINECSNumber | 233-038-3 |
RTECSNumber | GB5425000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.