Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chromium(III) Chloride Hexahydrate

Catalog No. 503081491 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-308-1491 500 g
50-308-1473 1 kg
50-308-1487 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Warning
Catalog No. 50-308-1491 Supplier Sigma Aldrich Supplier No. 27096500GF
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Chromium(III) chloride (CrCl3. 6H2O) hexahydrate can be used:As a source of chromium(II), which is in turn used as a catalyst in iodovinylation of aldehydes (Takai reaction).In the synthesis of chromium(III) complexes of tetradentate Schiff bases.In the preparation of Cr(III)-fatty acid complexes which can further be used as a catalyst for the synthesis of 1-monoacyIgIyceroIs.In combination with choline chloride as a deep eutectic solvent, which can be utilized as a catalyst for the esterification of formic and acetic acid with alcohols.

Specifications

CAS 10060-12-5
Density 1.76 g/mL at 25°C
Molecular Formula Cl3Cr -+ 6H2O
Linear Formula CrCl3 -+ 6H2O
MDL Number MFCD00149660
Quantity 500 g
Synonym Chromium trichloride hexahydrate; Hexaaquachromium(III) chloride
Molecular Weight (g/mol) 266.45 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.