Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium(III) Chloride Hexahydrate, MP Biomedicals

Catalog No. ICN19403380 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19403380 100 g
ICN19403390 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19403380 Supplier MP Biomedicals, Inc Supplier No. 0219403380
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10060-12-5
Molecular Formula Cl3CrH12O6
Molecular Weight (g/mol) 266.44
MDL Number MFCD00149660
InChI Key LJAOOBNHPFKCDR-UHFFFAOYSA-K
Synonym chromium iii chloride hexahydrate, chromic chloride hexahydrate, hexaaquachromium chloride, chromium trichloride hexahydrate, trichlorochromium hexahydrate, chromium chloride hexahydrate, hexaaquachromium iii chloride, chrom iii-chlorid, chromium 3+ chloride hexahydrate, chlorid chromity hexahydrat czech
PubChem CID 104957
ChEBI CHEBI:53442
IUPAC Name chromium(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3]

Specifications

Quantity 100 g
Formula Weight 266.4
Chemical Name or Material Chromium(III) Chloride Hexahydrate
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.