Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium(III) chloride hexahydrate, Puratronic™, 99.998% (metals basis)

Catalog No. p-7048492
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1068606 5 g
AA1068614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1068606 Supplier Thermo Scientific Chemicals Supplier No. 01068606
Only null left
Add to Cart

CAS: 10060-12-5 | Cl3CrH12O6 | 266.44 g/mol

Chromium(III) chloride hexahydrate is used as an organic synthesis catalyst and as an additive to intravenous solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10060-12-5
Molecular Formula Cl3CrH12O6
Molecular Weight (g/mol) 266.44
MDL Number MFCD00149660
InChI Key LJAOOBNHPFKCDR-UHFFFAOYSA-K
Synonym chromium iii chloride hexahydrate, chromic chloride hexahydrate, hexaaquachromium chloride, chromium trichloride hexahydrate, trichlorochromium hexahydrate, chromium chloride hexahydrate, hexaaquachromium iii chloride, chrom iii-chlorid, chromium 3+ chloride hexahydrate, chlorid chromity hexahydrat czech
PubChem CID 104957
ChEBI CHEBI:53442
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3]

Specifications

Melting Point 83°C
Color Green
Physical Form Crystalline Aggregates
Quantity 5 g
Linear Formula CrCl3·6H2O
UN Number UN3260
Merck Index 14,2222
Solubility Information Soluble in water and alcohol. Insoluble in diethyl ether.
IUPAC Name chromium(3+) hexahydrate trichloride
Formula Weight 266.45 (158.35 Anhydrous)
Percent Purity 99.998%
Grade Puratronic
Odor Odorless
Density 1.835 g/mL
Assay (metals basis)
Chemical Name or Material Chromium(III) chloride hexahydrate
Show More Show Less
Hazard Category H290-H302-H317
Hazard Statement GHS H Statement
H334-H314-H318-H302-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P234-P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P390-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 233-038-3
RTECSNumber GB5450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.