Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium(III) fluoride hydrate, Thermo Scientific™

Catalog No. p-7048979
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA12339A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA12339A1 Supplier Thermo Scientific Chemicals Supplier No. 12339A1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123333-98-2
Molecular Formula CrF3
Molecular Weight (g/mol) 108.99
MDL Number MFCD00016045
InChI Key FTBATIJJKIIOTP-UHFFFAOYSA-K
IUPAC Name chromium(3+) trifluoride
SMILES [F-].[F-].[F-].[Cr+3]

Specifications

Melting Point >1000°C
Color Green
Physical Form Crystalline
Quantity 1 kg
UN Number UN1756
Merck Index 14,2223
Solubility Information Insoluble in water.
Formula Weight 108.99 (Anhydrous)
Sensitivity Hygroscopic
Chemical Name or Material Chromium(III) fluoride hydrate
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHROMIC FLUORIDE, SOLID
EINECSNumber 232-137-9
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.