Learn More
Strem, An Ascensus Company CAS# 7789-02-8. 100g. Chromium(III) nitrate nonahydrate, 99%. MFCD00149659
SDP
Catalog No. 5090114535
Encompass
CAS# 7789-02-8. 100g. Chromium(III) nitrate nonahydrate, 99%. MFCD00149659. Molecular Weight: 238.06 (400.15). Molecular Formula: Cr(NO3)3∙9H2O. Color/form: greenish-black xtl. Strem# 93-2407. http://www.strem.com/catalog/v/93-2407/
Catalog No. 50-901-14535 Supplier Strem, An Ascensus Company Supplier No. 932407100G
CAS# 7789-02-8. 100g. Chromium(III) nitrate nonahydrate, 99%. MFCD00149659. Molecular Weight: 238.06 (400.15). Molecular Formula: Cr(NO3)3∙9H2O. Color/form: greenish-black xtl. Strem# 93-2407. http://www.strem.com/catalog/v/93-2407/