Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Chromium(III) oxide | 1308-38-9 | 99.9% | 151.99 g/mol | Cr2O3 | 100 G
SDP

Catalog No. 5000208451
Encompass_Preferred

Chromium(III) oxide (Cr2O3) is a high-purity inorganic solid used as a refractory material and pigment. It offers a high melting point, excellent oxidation resistance, and thermal stability for applications requiring durable, oxidation-resistant oxide materials.

  • High purity (99.9% trace metals basis) for consistent performance.
  • High melting point and oxidation resistance suitable for refractory applications.
  • Light green to green solid appearance for pigment and ceramic use.
  • Chemical formula Cr2O3 and molecular weight 151.99 g/mol for identification and calculations.
  • CAS number 1308-38-9 for unambiguous substance identification.

Catalog No. 50-002-08451 Supplier Medchemexpress LLC Supplier No. HYW088074A100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chromium(III) oxide (Cr2O3) is a high-purity inorganic solid used as a refractory material and pigment. It offers a high melting point, excellent oxidation resistance, and thermal stability for applications requiring durable, oxidation-resistant oxide materials.

  • High purity (99.9% trace metals basis) for consistent performance.
  • High melting point and oxidation resistance suitable for refractory applications.
  • Light green to green solid appearance for pigment and ceramic use.
  • Chemical formula Cr2O3 and molecular weight 151.99 g/mol for identification and calculations.
  • CAS number 1308-38-9 for unambiguous substance identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.