Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Chromium(III) oxide | 1308-38-9 | MFCD00010949 | 99.9% | 151.99 g/mol | Cr2O3 | 25 G
SDP

Catalog No. 5000208839
Encompass_Preferred

Chromium(III) oxide (Cr2O3) is an inorganic oxide used as a refractory material, pigment, and technical reagent; it is valued for a high melting point and oxidation resistance and is supplied at trace-metal grade for research and technical applications.

  • chemical formula: Cr2O3
  • purity: 99.9% trace metals basis
  • appearance: light green to green solid
  • molecular weight: 151.99 g/mol
  • cas number: 1308-38-9
  • storage: store at room temperature, keep dry and cool
  • intended use: for research use only; not for human use

Catalog No. 50-002-08839 Supplier Medchemexpress LLC Supplier No. 25GHYW088074A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chromium(III) oxide (Cr2O3) is an inorganic oxide used as a refractory material, pigment, and technical reagent; it is valued for a high melting point and oxidation resistance and is supplied at trace-metal grade for research and technical applications.

  • chemical formula: Cr2O3
  • purity: 99.9% trace metals basis
  • appearance: light green to green solid
  • molecular weight: 151.99 g/mol
  • cas number: 1308-38-9
  • storage: store at room temperature, keep dry and cool
  • intended use: for research use only; not for human use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.