Learn More
Medchemexpress LLC Chromium(III) oxide | 1308-38-9 | MFCD00010949 | 99.9% | 151.99 g/mol | Cr2O3 | 50 G
SDP
Catalog No. 5000208289
Encompass_Preferred
Chromium(III) oxide (Cr2O3) is a high-purity inorganic refractory powder (99.9% trace metals basis) with a high melting point and strong oxidation resistance, used in high-temperature ceramics, coatings, pigments, and research applications requiring stable chromium(III) oxide.
  • High purity (99.9% trace metals basis).
  • Inorganic refractory powder with high melting point and oxidation resistance.
  • Chemical formula: Cr2O3; molecular weight approximately 151.99 g/mol.
  • Common applications include ceramics, coatings, pigments, and specialized research.
  • Available in laboratory pack sizes (for example, 50 g).
Catalog No. 50-002-08289 Supplier Medchemexpress LLC Supplier No. HYW088074A50G
Chromium(III) oxide (Cr2O3) is a high-purity inorganic refractory powder (99.9% trace metals basis) with a high melting point and strong oxidation resistance, used in high-temperature ceramics, coatings, pigments, and research applications requiring stable chromium(III) oxide.
  • High purity (99.9% trace metals basis).
  • Inorganic refractory powder with high melting point and oxidation resistance.
  • Chemical formula: Cr2O3; molecular weight approximately 151.99 g/mol.
  • Common applications include ceramics, coatings, pigments, and specialized research.
  • Available in laboratory pack sizes (for example, 50 g).