Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium(III) phosphate tetrahydrate

Catalog No. AA2411018
Click to view available options
Quantity:
10 g
50 g

For Use As a Catalyst, Pigment | CAS: 10101-59-4 | CrH8O8P | 219.026 g/mol

It is used as an additive in drugs, anti-corrosive pigments, catalysts and polymer manufacture, is also used to catalyze cation exchange in sorption reactions, as a laminated structure to dampen vibration and noise in a motor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10101-59-4
Molecular Formula CrH8O8P
Molecular Weight (g/mol) 219.026
MDL Number MFCD00016049
InChI Key BVZHAJJRIRDSTG-UHFFFAOYSA-K
Synonym chromium iii phosphate tetrahydrate, cr.po4.4h2o, chromium 3+ ion tetrahydrate phosphate, chromium 3+ phosphate-water 1/1/4, chromium 3+ tetrahydrate phosphate
PubChem CID 53471880
IUPAC Name chromium(3+);phosphate;tetrahydrate
SMILES O.O.O.O.[O-]P(=O)([O-])[O-].[Cr+3]

Specifications

Physical Form Powder
Quantity 50 g
Solubility Information Soluble in acids. Insoluble in water.
Formula Weight 219.04 (146.97 Anhydrous)
Appearance Dark powder
Density 2.26
Chemical Name or Material Chromium(III) phosphate tetrahydrate
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 232-141-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.