Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Chromium(III) potassium sulfate dodecahydrate | 7788-99-0 | ≥98.0% | 499.40 g·mol⁻¹ | KCr(SO4)2·12H2O | 100 G
SDP

Catalog No. 5000299588
Encompass_Preferred

Chromium(III) potassium sulfate dodecahydrate is an inorganic salt used as a laboratory reagent and microscopic fixative. It is supplied at high purity for analytical and histological applications and is characterized by its defined dodecahydrate form and solubility in aqueous media.

  • Inorganic chromium(III) salt with defined hydration (dodecahydrate).
  • High purity (≥98.0%) suitable for laboratory and analytical use.
  • Molecular weight 499.40 g·mol⁻¹; formula KCr(SO4)2·12H2O.
  • Soluble in water and forms stable aqueous solutions.
  • Commonly used as a fixative in microscopy and in analytical chemistry.

Catalog No. 50-002-99588 Supplier Medchemexpress LLC Supplier No. HYW019950100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chromium(III) potassium sulfate dodecahydrate is an inorganic salt used as a laboratory reagent and microscopic fixative. It is supplied at high purity for analytical and histological applications and is characterized by its defined dodecahydrate form and solubility in aqueous media.

  • Inorganic chromium(III) salt with defined hydration (dodecahydrate).
  • High purity (≥98.0%) suitable for laboratory and analytical use.
  • Molecular weight 499.40 g·mol⁻¹; formula KCr(SO4)2·12H2O.
  • Soluble in water and forms stable aqueous solutions.
  • Commonly used as a fixative in microscopy and in analytical chemistry.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.