Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Chromium(III) potassium sulfate dodecahydrate | 7788-99-0 | MFCD00149917 | >98.0% | 499.40 g/mol | KCr(SO4)2·12H2O | 25 G
SDP

Catalog No. 5000303680
Encompass_Preferred

Chromium(III) potassium sulfate dodecahydrate (CAS 7788-99-0) is an inorganic salt, commonly called chrome alum, supplied as a pale purple crystalline solid. Offered as a ≥98.0% purity reagent, it serves as a stable source of chromium(III) for laboratory synthesis, analytical procedures, and industrial uses such as tanning and photographic film hardening.

  • High purity reagent (≥98.0%).
  • Pale purple crystalline solid.
  • Molecular formula KCr(SO4)2·12H2O; molar mass ~499.40 g/mol.
  • Provides a reliable source of chromium(III) for synthesis and analysis.
  • Applicable in tanning and photographic film hardening processes.

Catalog No. 50-003-03680 Supplier Medchemexpress LLC Supplier No. 25GHYW019950
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chromium(III) potassium sulfate dodecahydrate (CAS 7788-99-0) is an inorganic salt, commonly called chrome alum, supplied as a pale purple crystalline solid. Offered as a ≥98.0% purity reagent, it serves as a stable source of chromium(III) for laboratory synthesis, analytical procedures, and industrial uses such as tanning and photographic film hardening.

  • High purity reagent (≥98.0%).
  • Pale purple crystalline solid.
  • Molecular formula KCr(SO4)2·12H2O; molar mass ~499.40 g/mol.
  • Provides a reliable source of chromium(III) for synthesis and analysis.
  • Applicable in tanning and photographic film hardening processes.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.