Learn More
Medchemexpress LLC Chromium(III) potassium sulfate dodecahydrate | 7788-99-0 | MFCD00149917 | ≥98.0% | 499.40 | KCr(SO4)2·12H2O | 500 G
SDP
Catalog No. 5000301707
Encompass_Preferred
Chromium(III) potassium sulfate dodecahydrate (≥98%) is a research-grade inorganic reagent commonly used as a fixative in microscopic analysis and in analytical or preparative laboratory applications. It is supplied as a pale purple crystalline solid with formula KCr(SO4)2·12H2O and molecular weight 499.40.
Catalog No. 50-003-01707 Supplier Medchemexpress LLC Supplier No. HYW019950500G
Chromium(III) potassium sulfate dodecahydrate (≥98%) is a research-grade inorganic reagent commonly used as a fixative in microscopic analysis and in analytical or preparative laboratory applications. It is supplied as a pale purple crystalline solid with formula KCr(SO4)2·12H2O and molecular weight 499.40.