Learn More
Medchemexpress LLC Chromium(III) potassium sulfate dodecahydrate | 7788-99-0 | MFCD00149917 | 99.0-102.0% | 499.40 g·mol⁻¹ | KCr(SO4)2 · 12H2O | 1 KG
SDP
Catalog No. 5000299589
Encompass_Preferred
Chromium(III) potassium sulfate dodecahydrate is an inorganic crystalline salt (KCr(SO4)2·12H2O) used as a laboratory reagent and fixing agent. It is supplied for analytical and research use, and is commonly used to prepare chromium standard solutions and as a hardening/fixing agent in photographic and tanning processes.
  • High reagent purity suitable for analytical applications.
  • Dodecahydrate crystalline form with predictable stoichiometry.
  • Used to prepare chromium standard solutions.
  • Effective as a photographic film hardener and tanning agent.
  • Compatible with iodometric assay methods for chromium analysis.
Catalog No. 50-002-99589 Supplier Medchemexpress LLC Supplier No. HYW0199501KG
Chromium(III) potassium sulfate dodecahydrate is an inorganic crystalline salt (KCr(SO4)2·12H2O) used as a laboratory reagent and fixing agent. It is supplied for analytical and research use, and is commonly used to prepare chromium standard solutions and as a hardening/fixing agent in photographic and tanning processes.
  • High reagent purity suitable for analytical applications.
  • Dodecahydrate crystalline form with predictable stoichiometry.
  • Used to prepare chromium standard solutions.
  • Effective as a photographic film hardener and tanning agent.
  • Compatible with iodometric assay methods for chromium analysis.