Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chromium(III) sulfate hydrate

Catalog No. 342432100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-243-2100G 100 g
34-243-2500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 34-243-2100G Supplier Sigma Aldrich Supplier No. 342432100G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Chromium(III) sulfate hydrate

Specifications

Melting Point 100°C
CAS 15244-38-9
Quantity 100 g
Molecular Formula Cr2O12S3 -+ xH2O
Linear Formula Cr2(SO4)3 -+ xH2O
MDL Number MFCD00150215
Synonym Chromic sulfate hydrate; Chromium trisulfate
Molecular Weight (g/mol) 392.18
Density 1.7 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.