Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium(III) telluride, 99.5% (metals basis)

Catalog No. AA4020614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4020614 25 g
AA4020606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4020614 Supplier Thermo Scientific Chemicals Supplier No. 04020614
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 12053-39-3 | Cr2Te3 | 486.79 g/mol

Chromium telluride is used in a number of industrial applications including infrared detection and imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12053-39-3
Molecular Formula Cr2Te3
Molecular Weight (g/mol) 486.79
MDL Number MFCD00016040
InChI Key PDJHBRMODJKXLQ-UHFFFAOYSA-N
Synonym tellanylidene tellanylidenechromio tellanyl chromium, chromium iii telluride, chromium telluride, chromium iii telluride,-325 mesh
PubChem CID 6381587
IUPAC Name tellanylidenechromium;tellurium
SMILES [Te].[Cr]=[Te].[Cr]=[Te]

Specifications

Melting Point ∼1300°C
Color Gray to Black
Physical Form Powder
Quantity 25 g
Odor Odorless
Density 7.0 g/mL
Solubility Information Insoluble in water.
Formula Weight 486.79
Percent Purity 99.5%
Assay (metals basis)
Chemical Name or Material Chromium(III) telluride
Show More Show Less
Hazard Category H302+H332-H317-H350
Hazard Statement GHS H Statement
H301-H350-H332-H317
Toxic if swallowed.
May cause cancer.
Harmful if inhaled.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P272-P280g-P281-P301+P312-P302+P352-P304+P340-P308+P313-P330-P333+P313-P363-P501c
EINECSNumber 235-003-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.