Learn More
Costech Analytical Technologies Inc CHROMIUM OXIDE GRAINS 50GR/BTL
Catalog No. NC9485349
Encompass
Chromium oxide catalyst, 50 gm, Catalysts and Reagent for: N, N/C and CHN
Catalog No. NC9485349 Supplier Costech Analytical Technologies Inc Supplier No. 011001
Chromium oxide catalyst, 50 gm, Catalysts and Reagent for: N, N/C and CHN