Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Chromium picolinate | 14639-25-9 | MFCD00068715 | 99.8% | 418.3 g/mol | C18H12CrN3O6 | 10MM 1ML
SDP

Catalog No. 5000235064
Encompass_Preferred

Chromium picolinate is a coordination complex of chromium(III) and picolinic acid used as an analytical reagent in biochemical and cellular research, including studies of insulin signaling. It is provided as a ready-to-use 10 mM solution in DMSO (1 mL) or as solid formats for reconstitution, and is accompanied by safety and handling documentation.

  • High purity (~99.8%) suitable for analytical and research applications.
  • Available as a 10 mM solution in DMSO for immediate use.
  • Also available as solid formats for reconstitution or bulk use.
  • Chemical formula C18H12CrN3O6 and molecular weight 418.3 g/mol.
  • Supplied with safety data sheet and technical documentation for safe handling.

Catalog No. 50-002-35064 Supplier Medchemexpress LLC Supplier No. HY12558810MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chromium picolinate is a coordination complex of chromium(III) and picolinic acid used as an analytical reagent in biochemical and cellular research, including studies of insulin signaling. It is provided as a ready-to-use 10 mM solution in DMSO (1 mL) or as solid formats for reconstitution, and is accompanied by safety and handling documentation.

  • High purity (~99.8%) suitable for analytical and research applications.
  • Available as a 10 mM solution in DMSO for immediate use.
  • Also available as solid formats for reconstitution or bulk use.
  • Chemical formula C18H12CrN3O6 and molecular weight 418.3 g/mol.
  • Supplied with safety data sheet and technical documentation for safe handling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.