Learn More
Selleck Chemical LLC Chromium picolinate
SDP
Catalog No. 5000004681
Encompass_Preferred
Chromium picolinate is a coordination complex consisting of chromium(III) and picolinic acid It is used as a nutritional supplement for optimal insulin function in patients with Type 2 diabetes or promotion of weight loss
Catalog No. 50-000-04681 Supplier Selleck Chemical LLC Supplier No. S644325MG
Chromium picolinate is a coordination complex consisting of chromium(III) and picolinic acid It is used as a nutritional supplement for optimal insulin function in patients with Type 2 diabetes or promotion of weight loss