Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Chromium Picolinate
SDP

Catalog No. 14185491 Shop All Medisca Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-185-491 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-185-491 Supplier Medisca Supplier No. 117404
Add to Cart
Add to Cart

Chromium Picolinate comes as either an odorless powder or solid, that is pink (lavender) to red in color. It is insoluble in water, but soluble in polar solvents.

Chromium Picolinate comes as either an odorless powder or solid, that is pink (lavender) to red in color. It is insoluble in water, but soluble in polar solvents.

Alternative name(s): Chromium tripicolinate

Specifications

Chemical Name or Material Chromium Picolinate
Packaging Jar, 100mL, White, HDPE, Round, 38-400, TE Pano Neck
CAS 14639-25-9
Type Drug Standard
Molecular Weight (g/mol) 418.3038
Quantity 25 g
Percent Purity 12.06 to 12.56 %
Grade Ungraded
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.