Learn More
SPEX CERTIPREP LLC 3P CHROMIUM PLUS 6 STANDARD 100ML
Supplier:  SPEX CERTIPREP LLC 3P 1000127100
Encompass_Preferred
(HPS by Zeptometrix) Chromium, Source: K2Cr2O7 as Cr+6, Purity: 99.998%, Matrix: H2O, 100mL
Catalog No. NC9865538
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.