Learn More
Bioworld Chromium Potassium Sulfate, Dodecahydrate, 50 g
Catalog No. 50255704
Encompass_Preferred
Specifications:ACS ReagentSynonium: Chrome alum, Chromium (III) Potassium sulfate dodecahydrateAppearance: Crystalline or chunksDensity : 1.83 g/mlMelting Point: 89 ºCRTECS Number: GB6850000
Catalog No. 50-255-704 Supplier Bioworld Supplier No. 403001542
Specifications:ACS ReagentSynonium: Chrome alum, Chromium (III) Potassium sulfate dodecahydrateAppearance: Crystalline or chunksDensity : 1.83 g/mlMelting Point: 89 ºCRTECS Number: GB6850000