Learn More
Strem, An Ascensus Company CAS# 7440-47-3. 100g. Chromium powder (99%). MFCD00010944
SDP
Catalog No. 5090114576
Encompass
CAS# 7440-47-3. 100g. Chromium powder (99%). MFCD00010944. Molecular Weight: 52.01. Molecular Formula: Cr. Color/form: -100 mesh Strem# 93-2458. http://www.strem.com/catalog/v/93-2458/
Catalog No. 50-901-14576 Supplier Strem, An Ascensus Company Supplier No. 932458100G
CAS# 7440-47-3. 100g. Chromium powder (99%). MFCD00010944. Molecular Weight: 52.01. Molecular Formula: Cr. Color/form: -100 mesh Strem# 93-2458. http://www.strem.com/catalog/v/93-2458/