Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical™

Catalog No. SC192100 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SC192-100 100 mL Poly Bottle
SC192-500 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. SC192-100
This item is not returnable. View return policy

For atomic absorption. (1mL = 1mg Cr) Solute: Potassium Dichromate Solvent: Distilled H2O

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-50-9 , 7732-18-5
Molecular Formula Cr2K2O7
Molecular Weight (g/mol) 294.182
MDL Number MFCD00010944
InChI Key KMUONIBRACKNSN-UHFFFAOYSA-N
PubChem CID 24502
ChEBI CHEBI:53444
IUPAC Name dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
SMILES [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]

Specifications

Packaging Poly Bottle
Vapor Pressure 14mmHg at 20°C
Density 1.0g/cm³
Physical Form Liquid
Concentration 990 to 1010 ppm
Color Orange
pH 7
Melting Point 0°C
Boiling Point 100°C
Quantity 100 mL
Grade Certified
Concentration or Composition (by Analyte or Components) 1000ppm ±1%
Chemical Name or Material Chromium, Reference Standard Solution, 1000ppm ±1%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.