Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium Standard, 1mL = 1mg Cr, 1000ppm CrO42-, Ricca Chemical
SDP

Catalog No. 21004 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Poly natural
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
2100-4 120 mL Poly natural
210016 500 mL Poly natural
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 2100-4 Supplier Ricca Chemical Company Supplier No. 21004

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

This item is not returnable. View return policy

Chemical Identifiers

CAS 7789-00-6 , 7732-18-5
Molecular Formula CrK2O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00011368
InChI Key XMXNVYPJWBTAHN-UHFFFAOYSA-N
PubChem CID 24597
ChEBI CHEBI:75249
IUPAC Name dipotassium dioxochromiumbis(olate)
SMILES [K+].[K+].[O-][Cr]([O-])(=O)=O

Specifications

Packaging Poly natural
Concentration or Composition Notes 1000ppm Cr
Equivalence 1 mL = 1 mg Cr
Physical Form Liquid
Color Yellow
Quantity 120 mL
Chemical Name or Material Chromium Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.