Learn More
Hach Company Chromium TNTplus Vial Test (0.03-1.00 mg/L Cr), 25 Tests
Catalog No. 501998143 Shop All Hach Company Products
Encompass
Chromium TNTplus chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime.For determination of Hexavalent and Total Chromium by the 1,5-Diphenylcarbohydrazide Method. Total chromium determination requires digestion.
Catalog No. 50-199-8143 Supplier Hach Company Supplier No. TNT854
Chromium TNTplus chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime.For determination of Hexavalent and Total Chromium by the 1,5-Diphenylcarbohydrazide Method. Total chromium determination requires digestion.