Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hach Company Hach Chromium Total Reagent Set, 0.01-0.70 mg/L Cr (MPN 2242500)

Catalog No. NC3304604 Shop All Hach Company Products
Encompass

Reagent set for determination of total chromium in water and wastewater using the alkaline hypobromite oxidation method (Method 8024). Supplies acid and sealed reagent powder pillows plus a method blank for colorimetric or spectrophotometric measurement.

  • Measurement range: 0.01-0.70 mg/L Cr (spectrophotometers); 0.01-0.60 mg/L Cr (colorimeters).
  • Intended method: Alkaline hypobromite oxidation (Method 8024).
  • Components included: 5.25 N sulfuric acid solution, reagent powder pillows for chromium analysis, and 100 mL method blank (MDB).
  • Package quantity: 100 reagent sets per pack.
  • Applications: Water and wastewater total chromium testing.
  • Notes: Procedure requires a boiling water bath; measure reagent blank for each new lot; review SDS and use appropriate PPE.

Catalog No. NC3304604 Supplier Hach Company Supplier No. 2242500
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Reagent set for determination of total chromium in water and wastewater using the alkaline hypobromite oxidation method (Method 8024). Supplies acid and sealed reagent powder pillows plus a method blank for colorimetric or spectrophotometric measurement.

  • Measurement range: 0.01-0.70 mg/L Cr (spectrophotometers); 0.01-0.60 mg/L Cr (colorimeters).
  • Intended method: Alkaline hypobromite oxidation (Method 8024).
  • Components included: 5.25 N sulfuric acid solution, reagent powder pillows for chromium analysis, and 100 mL method blank (MDB).
  • Package quantity: 100 reagent sets per pack.
  • Applications: Water and wastewater total chromium testing.
  • Notes: Procedure requires a boiling water bath; measure reagent blank for each new lot; review SDS and use appropriate PPE.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.