Learn More
Specifications
Specifications
| Color | Purple to Red |
| CAS Min % | 98.0 |
| Physical Form | Crystalline Solid |
| CAS Max % | 100.0 |
| Quantity | 500 g |
| Packaging | White Poly Wide Mouth Bottle |
| Percent Purity | ≥ 98% |
| Grade | ACS |
| Chemical Name or Material | Chromium Trioxide |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
