Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium Trioxide (Flakes/Technical), Fisher Chemical™

Catalog No. A98500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A98-500 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. A98-500

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1333-82-0
Molecular Formula CrO3
Molecular Weight (g/mol) 99.99
MDL Number MFCD00010952
InChI Key WGLPBDUCMAPZCE-UHFFFAOYSA-N
Synonym chromium trioxide, chromium vi oxide, monochromium trioxide, chromtrioxid, chromic trioxide, chromsaeureanhydrid, anhydride chromique, chromium 6+ oxide, chromium vi trioxide, chromium oxide
PubChem CID 14915
ChEBI CHEBI:48240
IUPAC Name trioxochromium
SMILES O=[Cr](=O)=O

Specifications

Boiling Point 250°C
Melting Point 196°C
Color Violet
pH 1
Physical Form Solid
Quantity 500 g
UN Number UN 1463
Formula Weight 99.99
Grade Technical
Identification Pass Test
Packaging Glass Bottle
Density 2.700g/cm³
Chemical Name or Material Chromium Trioxide
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Explosive when mixed with combustible material. May be fatal if inhaled. Causes severe burns by all exposure routes. Toxic in contact with skin and if swallowed. May cause sensitization by inhalation and skin contact. Danger of serious damage to health by prolonged exposure. May cause heritable genetic damage. Possible risk of impaired fertility. Hygroscopic. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Wear personal protective equipment. Use only under a chemical fume hood. Use only under a chemical fume hood. Keep away from clothing and other combustible materials.

NFPA

Health:3
Flammability:0
Instability:1
DOTInformation DOT Class 5.1, : Oxidizer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.