Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium(VI) AA Standard, 1mL = 1mg Cr (1,000ppm Cr) in H2O, Ricca Chemical

Click to view available options
Quantity:
100mL
500mL
Packaging:
Poly natural
7778-50-9, 7732-18-5
Cr2K2O7
294.182
KMUONIBRACKNSN-UHFFFAOYSA-N
24502
CHEBI:53444
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
7778-50-9, 7732-18-5
Cr2K2O7
294.182
KMUONIBRACKNSN-UHFFFAOYSA-N
24502
CHEBI:53444
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
CAS Min % 98.0, 0.28
CAS Max % 100.0, 0.29
Physical Form Liquid
Packaging Poly natural
Equivalence 1 mL = 1 mg Cr
Color Yellow
Quantity 100mL
Concentration or Composition Notes 1,000ppm Cr
Chemical Name or Material Chromium(VI) AA Standard