Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium(VI) Oxide, ≥98.0%, ACS Reagent, Solstice

Catalog No. 60045440 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-440 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-045-440 Supplier Solstice Supplier No. 236470100G

missing translation for 'validMainframeMsg'

ACS Reagent,≥98.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1333-82-0
Molecular Formula CrO3
Molecular Weight (g/mol) 99.99
MDL Number MFCD00010952
InChI Key WGLPBDUCMAPZCE-UHFFFAOYSA-N
Synonym chromium trioxide, chromium vi oxide, monochromium trioxide, chromtrioxid, chromic trioxide, chromsaeureanhydrid, anhydride chromique, chromium 6+ oxide, chromium vi trioxide, chromium oxide
PubChem CID 14915
ChEBI CHEBI:48240
IUPAC Name trioxochromium
SMILES O=[Cr](=O)=O

Specifications

Melting Point 196°C
Color Violet
Quantity 100 g
UN Number UN1463
Formula Weight 99.99g/mol
Percent Purity ≥98.0%
Grade ACS Reagent
Packaging Glass Bottle
Appearance Powder
Density 2.7g/cm3
Chemical Name or Material Chromium(Vi) Oxide
Show More Show Less
Precautionary Statement H271-H301 + H311-H314-H317-H330-H334-H340-H350-H361f-H372-H410
DOTInformation 8(6.1)(5.1)/PGII
EINECSNumber 215-607-8
RTECSNumber GB6650000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.