Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium(VI) oxide, 98+%, ACS reagent

Catalog No. AC405230025
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC405230025 2.5 kg Glass bottle
AC405230010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC405230025 Supplier Thermo Scientific Chemicals Supplier No. 405230025
Add to Cart
This item is not returnable. View return policy

CAS: 1333-82-0 | CrO3 | 99.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1333-82-0
Molecular Formula CrO3
Molecular Weight (g/mol) 99.99
MDL Number MFCD00010952
InChI Key WGLPBDUCMAPZCE-UHFFFAOYSA-N
Synonym chromium trioxide, chromium vi oxide, monochromium trioxide, chromtrioxid, chromic trioxide, chromsaeureanhydrid, anhydride chromique, chromium 6+ oxide, chromium vi trioxide, chromium oxide
PubChem CID 14915
ChEBI CHEBI:48240
IUPAC Name trioxochromium
SMILES O=[Cr](=O)=O

Specifications

Melting Point 196.0°C
Color Brown-Red to Purple
Physical Form Solid
Quantity 2.5 kg
Assay Percent Range 98+%
Linear Formula CrO3
Fieser 01,144; 02,72; 03,54; 04,96; 05,140; 07,70; 09,115; 10,99; 13,338; 14,60
Merck Index 15, 2239
Solubility Information Solubility in water: 1660g/L (20°C). Other solubilities: soluble in sulfuric acid, soluble in alcohol and mineral acids
Formula Weight 99.99
Percent Purity ≥98%
Grade ACS Reagent
Packaging Glass bottle
Specific Gravity 2.7
Density 2.7000g/mL
Chemical Name or Material Chromium(VI) oxide, 98+%
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Fatal if inhaled. Causes damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause an allergic skin reaction. Suspected of damaging fertility. Very toxic to aquatic life with long lasting effects. May cause genetic defects. May cause cancer. May cause respiratory irritation. Fatal in contact with skin. May cause fire or explosion; strong oxidizer. Toxic if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Gently wash with plenty of soap and water. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Do not breathe dust/fume/gas/mist/vapors/spray. Avoid release to the environment.
EINECSNumber 215-607-8
RTECSNumber GB6650000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.