Learn More
Cayman Chemical Chroman 1 (hydrochloride hydrate), Purity: ≥98%, 10 mg
SDP
Catalog No. 502910227
Encompass_Preferred
A ROCK2 inhibitor (IC50 = 50s = 1,740 and 127 nM, respectively); inhibits phosphorylation of MLC in a cell-based assay (IC50 = <4 nM)
Catalog No. 50-291-0227 Supplier Cayman Chemical Supplier No. 3468210 MG
A ROCK2 inhibitor (IC50 = 50s = 1,740 and 127 nM, respectively); inhibits phosphorylation of MLC in a cell-based assay (IC50 = <4 nM)