Learn More
Cayman Chemical Chromnol 293B 50mg
SDP
Catalog No. 502983968
Encompass_Preferred
An IKs and ICTFR blocker (IC50s = 6.89 and 19 μM in Xenopus oocytes expressing rat IKs and CTFR, respectively); has no activity at rat Kv1.1 or Kir2.1 channels at a concentration of 30 μM; increases the rate and extent of IKs currents in guinea pig ventricular cells in a dose-dependent manner
Catalog No. 50-298-3968 Supplier Cayman Chemical Supplier No. 1999350 MG
An IKs and ICTFR blocker (IC50s = 6.89 and 19 μM in Xenopus oocytes expressing rat IKs and CTFR, respectively); has no activity at rat Kv1.1 or Kir2.1 channels at a concentration of 30 μM; increases the rate and extent of IKs currents in guinea pig ventricular cells in a dose-dependent manner