Learn More
Apexbio Technology LLC Chromocarb 4940-39-0 50mg
SDP
Catalog No. 502240769
Encompass_Preferred
Chromocarb is a synthetic small-molecule compound designed to improve endothelial cellular function reduce endothelial injury and suppress vascular inflammation signaling pathways Chromocarb exerts its biological activity primarily through modulation of molecular pathways associated with endothelial cell damage and inflammatory responses in the vasculature It is utilized in research to investigate the molecular mechanisms underlying vascular pathology including models of tissue injury ischemia-reperfusion injury and vascular complications related to diabetes Based on these pharmacological properties Chromocarb holds research potential in studying endothelial protection and vascular inflammatory diseases
This item is not returnable. View return policy
Catalog No. 50-224-0769 Supplier Apexbio Technology LLC Supplier No. B170450
Chromocarb is a synthetic small-molecule compound designed to improve endothelial cellular function reduce endothelial injury and suppress vascular inflammation signaling pathways Chromocarb exerts its biological activity primarily through modulation of molecular pathways associated with endothelial cell damage and inflammatory responses in the vasculature It is utilized in research to investigate the molecular mechanisms underlying vascular pathology including models of tissue injury ischemia-reperfusion injury and vascular complications related to diabetes Based on these pharmacological properties Chromocarb holds research potential in studying endothelial protection and vascular inflammatory diseases