Learn More
Genscript Corporation ToxinSensor™ Chromogenic LAL Endotoxin Assay Kit
Catalog No. NC2217212
Encompass
Genscript ToxinSensor™ Chromogenic LAL Endotoxin Assay Kit is designed to be a quantitative In Vitro end-point endotoxin test for human and animal parenteral drugs, biological products, and medical devices.The system is not intended for detection of endotoxin in a licensed reagent, clinical samples or the diagnosis of human disease. A measurable endotoxin concentration range from 0.01 to 1 EU/ml can be achieved. Lyophilized Amebocyte Lysate reagent is made from amebocyte lysate from the horseshoe crab (Tachypleus tridentatus).
This item is not returnable. View return policy
Catalog No. NC2217212 Supplier Genscript Corporation Supplier No. L00350C16
 Restricted Item
Add to Cart
Add to Cart
Genscript ToxinSensor™ Chromogenic LAL Endotoxin Assay Kit is designed to be a quantitative In Vitro end-point endotoxin test for human and animal parenteral drugs, biological products, and medical devices.The system is not intended for detection of endotoxin in a licensed reagent, clinical samples or the diagnosis of human disease. A measurable endotoxin concentration range from 0.01 to 1 EU/ml can be achieved. Lyophilized Amebocyte Lysate reagent is made from amebocyte lysate from the horseshoe crab (Tachypleus tridentatus).