Learn More
Genscript Corporation ToxinSensor™ Chromogenic LAL Endotoxin Assay Kit
Supplier:  Genscript Corporation L00350
Encompass
Genscript ToxinSensor™ Chromogenic LAL Endotoxin Assay Kit is designed to be a quantitative In Vitro end-point endotoxin test for human and animal parenteral drugs, biological products, and medical devices.The system is not intended for detection of endotoxin in a licensed reagent, clinical samples or the diagnosis of human disease. A measurable endotoxin concentration range from 0.01 to 1 EU/ml can be achieved. Lyophilized Amebocyte Lysate reagent is made from amebocyte lysate from the horseshoe crab (Tachypleus tridentatus).
Catalog No. 50-274-479